Assay Details
Gene Family ID: | MIPF0000405, mir-673 |
Mature miRNA Sequence: | CUCACAGCUCUGGUCCUUGGAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-673-5p | MIMAT0003739 |
Stem-loop Details
Mouse

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | mmu-miR-673-5p |
---|---|
miRBase Accession Number: | MI0004601 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CUCACAGCUCUGGUCCUUGGAG |
Species: | Mouse |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000405, mir-673 |
Mature miRNA Sequence: | CUCACAGCUCUGGUCCUUGGAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-673-5p | MIMAT0003739 |