Assay Details
Amplicon Length: | 88 |
Chromosome Location: | Chr. 12 - 109571789 - 109571789 [-] on 109571789 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-673-5p | MIMAT0003739 |
| Mouse | mmu-miR-673-3p | MIMAT0004824 |
Stem-loop Details
Mouse

Genomic Map