Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | CAAAGCGCUUCUCUUUAGAGUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-518c-3p | MIMAT0002848 |
| Gorilla gorilla | ggo-miR-518c-3p | MIMAT0036422 |
| Pan troglodytes | ptr-miR-518c | MIMAT0008188 |
| Pongo pygmaeus | ppy-miR-518c | MIMAT0015954 |
Stem-loop Details
Human
Gorilla gorilla
Pan troglodytes
Pongo pygmaeus

Genomic Map