Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | CUCUAGAGGGAAGCACUUUCUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-518f-5p | MIMAT0002841 |
| Gorilla gorilla | ggo-miR-518f-5p | MIMAT0036416 |
Stem-loop Details
Human
Gorilla gorilla

Genomic Map