Assay Details
Gene Family ID: | MIPF0000147, mir-325 |
Mature miRNA Sequence: | UUUAUUGAGCACCUCCUAUCAA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-325-3p | MIMAT0004640 |
Rat | rno-miR-325-3p | MIMAT0004639 |
Rabbit | ocu-miR-325-3p | MIMAT0048373 |
Stem-loop Details
Mouse
Rat
Rabbit
