Assay Details
Amplicon Length: | 67 |
Chromosome Location: | Chr. X - 76334527 - 76334527 [-] on 76334527 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-325-3p | MIMAT0004639 |
Rat | rno-miR-325-5p | MIMAT0000557 |
Stem-loop Details
Rat
