Assay Details
Gene Family ID: | MIPF0000071, mir-302 |
Mature miRNA Sequence: | ACUUUAACAUGGAGGCACUUGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-302d-5p | MIMAT0004685 |
| Callithrix jacchus | cja-miR-302d-5p | MIMAT0039589 |
| Dog | cfa-miR-302d | MIMAT0009858 |
| Rabbit | ocu-miR-302d-5p | MIMAT0036322 |
Stem-loop Details
Human
Callithrix jacchus
Dog
Rabbit

Genomic Map