Assay Details
Mature miRNA Sequence: | UGAGGUAGUAGUUAGAA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-1961 | MIMAT0009434 |
Stem-loop Details
Mouse

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | mmu-miR-1961 |
---|---|
miRBase Accession Number: | MI0009958 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UGAGGUAGUAGUUAGAA |
Species: | Mouse |
Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UGAGGUAGUAGUUAGAA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-1961 | MIMAT0009434 |