Assay Details
Amplicon Length: | 108 |
Chromosome Location: | Chr. 5 - 92788796 - 92788796 [-] on 92788796 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1961 | MIMAT0009434 |
Stem-loop Details
Mouse
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 108 |
Chromosome Location: | Chr. 5 - 92788796 - 92788796 [-] on 92788796 Build GRCm38 |
Species: | Mouse |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1961 | MIMAT0009434 |