Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | AAAAACCACAAUUACUUUUGCACCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548aa | MIMAT0018447 |
| Human | hsa-miR-548t-3p | MIMAT0022730 |
Stem-loop Details
Human

Genomic Map