Assay Details
Gene Family ID: | MIPF0000470, mir-549 |
Mature miRNA Sequence: | AGCUCAUCCAUAGUUGUCACUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-549a-5p | MIMAT0037328 |
| Nomascus leucogenys | nle-miR-549 | MIMAT0049377 |
| Pan paniscus | ppa-miR-549 | MIMAT0049096 |
Stem-loop Details
Human
Nomascus leucogenys
Pan paniscus

Genomic Map