Assay Details
Amplicon Length: | 66 |
Chromosome Location: | Chr. 7 - 19181768 - 19181768 [-] on 19181768 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-330-5p | MIMAT0004642 |
| Mouse | mmu-miR-330-3p | MIMAT0000569 |
Stem-loop Details
Mouse

Genomic Map