Assay Details
Gene Family ID: | MIPF0000200, mir-330 |
Mature miRNA Sequence: | UCUCUGGGCCUGUGUCUUAGGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-330-5p | MIMAT0004693 |
| Mouse | mmu-miR-330-5p | MIMAT0004642 |
| Rat | rno-miR-330-5p | MIMAT0004641 |
| Dog | cfa-miR-330 | MIMAT0009893 |
| Horse | eca-miR-330 | MIMAT0013012 |
| Pongo pygmaeus | ppy-miR-330-5p | MIMAT0015833 |
| View More Rows | ||
| Rhesus monkey | mml-miR-330-5p | MIMAT0006270 |
Stem-loop Details
Human
Mouse
Rat
Dog
Horse
Pongo pygmaeus
Rhesus monkey

Genomic Map