Assay Details
Amplicon Length: | 72 |
Chromosome Location: | Chr. 12 - 109563394 - 109563394 [-] on 109563394 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-770-5p | MIMAT0004822 |
| Mouse | mmu-miR-770-3p | MIMAT0003891 |
Stem-loop Details
Mouse

Genomic Map