Assay Details
Gene Family ID: | MIPF0000355, mir-770 |
Mature miRNA Sequence: | AGCACCACGUGUCUGGGCCACG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-770-5p | MIMAT0004822 |
| Rat | rno-miR-770-5p | MIMAT0005338 |
Stem-loop Details
Mouse
Rat

Genomic Map