Assay Details
Amplicon Length: | 78 |
Chromosome Location: | Chr. 5 - 111003945 - 111003945 [-] on 111003945 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-701-5p | MIMAT0003491 |
| Mouse | mmu-miR-701-3p | MIMAT0017257 |
Stem-loop Details
Mouse

Genomic Map