Assay Details
Mature miRNA Sequence: | UUAGCCGCUGAAAUAGAUGGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-701-5p | MIMAT0003491 |
Stem-loop Details
Mouse

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | mmu-miR-701-5p |
---|---|
Stem-loop Accession Number: | MI0004685 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UUAGCCGCUGAAAUAGAUGGA |
Species: | Mouse |
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | UUAGCCGCUGAAAUAGAUGGA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-701-5p | MIMAT0003491 |