Assay Details
Amplicon Length: | 81 |
Chromosome Location: | Chr. X - 66796500 - 66796500 [+] on 66796500 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-741-3p | MIMAT0004236 |
| Mouse | mmu-miR-741-5p | MIMAT0017262 |
Stem-loop Details
Mouse

Genomic Map