Assay Details
| Mature miRNA Sequence: | UGAGAGAUGCCAUUCUAUGUAGA | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Mouse | mmu-miR-741-3p | MIMAT0004236 | 
Stem-loop Details
Mouse
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mmu-miR-741-3p | 
|---|---|
| miRBase Accession Number: | MI0005205 | 
| miRBase Version: | v22.1 | 
| Mature miRNA Sequence: | UGAGAGAUGCCAUUCUAUGUAGA | 
| Species: | Mouse | 
| Product Type: | Ambion® Pre-miR™ miRNA Precursor | 
| Mature miRNA Sequence: | UGAGAGAUGCCAUUCUAUGUAGA | 
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Mouse | mmu-miR-741-3p | MIMAT0004236 | 
 Back To Top
 Back To Top