Assay Details
Amplicon Length: | 100 |
Chromosome Location: | Chr. 3 - 90070316 - 90070316 [+] on 90070316 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-190b-3p | MIMAT0017267 |
| Mouse | mmu-miR-190b-5p | MIMAT0004852 |
Stem-loop Details
Mouse

Genomic Map