Assay Details
Amplicon Length: | 74 |
Chromosome Location: | Chr. 12 - 109612212 - 109612212 [+] on 109612212 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1188-3p | MIMAT0017328 |
| Mouse | mmu-miR-1188-5p | MIMAT0005843 |
Stem-loop Details
Mouse

Genomic Map