Assay Details
Gene Family ID: | MIPF0001188, mir-1188 |
Mature miRNA Sequence: | UGGUGUGAGGUUGGGCCAGGA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-1188-5p | MIMAT0005843 |
Rat | rno-miR-1188-5p | MIMAT0017854 |
Stem-loop Details
Mouse
Rat
