Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAUCGUACAGGGUCAUCCACU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gorilla gorilla | ggo-miR-487b | MIMAT0024105 |
Stem-loop Details
Gorilla gorilla
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-487b (v9.2)
Rat: rno-miR-487b (v15)
Rat: rno-miR-487b (v15)

Genomic Map