Assay Details
Gene Family ID: | MIPF0000353, mir-802 |
Mature miRNA Sequence: | UCAGUAACAAAGAUUCAUCCUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-802-5p | MIMAT0004188 |
Pteropus alecto | pal-miR-802-5p | MIMAT0040179 |
Stem-loop Details
Mouse
Pteropus alecto

More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-802 (v17)
Rat: rno-miR-802 (v15)
Rat: rno-miR-802 (v15)
