Assay Details
Gene Family ID: | MIPF0000353, mir-802 |
Mature miRNA Sequence: | UCAGUAACAAAGAUUCAUCCUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-802-5p | MIMAT0004188 |
| Pteropus alecto | pal-miR-802-5p | MIMAT0040179 |
Stem-loop Details
Mouse
Pteropus alecto

Genomic Map