Assay Details
Gene Family ID: | MIPF0000024, mir-103 |
Mature miRNA Sequence: | AGCUUCUUUACAGUGUUGCCUUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-107-5p | MIMAT0017115 |
| Ophiophagus hannah | oha-miR-107-5p | MIMAT0036654 |
Stem-loop Details
Rat
Ophiophagus hannah

Genomic Map