Assay Details
Amplicon Length: | 73 |
Chromosome Location: | Chr. 1 - 253128090 - 253128090 [-] on 253128090 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-107-3p | MIMAT0000826 |
| Rat | rno-miR-107-5p | MIMAT0017115 |
Stem-loop Details
Rat

Genomic Map