Assay Details
Amplicon Length: | 69 |
Chromosome Location: | Chr. 6 - 133683218 - 133683218 [-] on 133683218 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-770-3p | MIMAT0017317 |
| Rat | rno-miR-770-5p | MIMAT0005338 |
Stem-loop Details
Rat

Genomic Map