Assay Details
Amplicon Length: | 62 |
Chromosome Location: | Chr. 1 - 80092532 - 80092532 [+] on 80092532 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-330-5p | MIMAT0004641 |
| Rat | rno-miR-330-3p | MIMAT0000568 |
Stem-loop Details
Rat

Genomic Map