Assay Details
Gene Family ID: | MIPF0000399, mir-872 |
Mature miRNA Sequence: | AAGGUUACUUGUUAGUUCAGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-872-5p | MIMAT0004934 |
| Rat | rno-miR-872-5p | MIMAT0005282 |
| Cricetulus griseus | cgr-miR-872-5p | MIMAT0024005 |
| Guinea pig | cpo-miR-872-5p | MIMAT0047389 |
| Horse | eca-miR-872 | MIMAT0013119 |
| Microcebus murinus | mmr-miR-872 | MIMAT0049152 |
| View More Rows | ||
| Rabbit | ocu-miR-872-5p | MIMAT0048571 |
Stem-loop Details
Mouse
Rat
Cricetulus griseus
Guinea pig
Horse
Microcebus murinus
Rabbit

Genomic Map