Assay Details
Amplicon Length: | 89 |
Chromosome Location: | Chr. 5 - 113658072 - 113658072 [+] on 113658072 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-872-3p | MIMAT0005283 |
Rat | rno-miR-872-5p | MIMAT0005282 |
Stem-loop Details
Rat
