Assay Details
Gene Family ID: | MIPF0000399, mir-872 |
Mature miRNA Sequence: | UGAACUAUUGCAGUAGCCUCCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-872-3p | MIMAT0004935 |
Rat | rno-miR-872-3p | MIMAT0005283 |
Stem-loop Details
Mouse
Rat
