Assay Details
Gene Family ID: | MIPF0000147, mir-325 |
Mature miRNA Sequence: | CCUAGUAGGUGCUCAGUAAGUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-325-5p | MIMAT0000558 |
| Rat | rno-miR-325-5p | MIMAT0000557 |
Stem-loop Details
Mouse
Rat

Genomic Map