Assay Details
Gene Family ID: | MIPF0000139, mir-500 |
Mature miRNA Sequence: | AAUGCACCCGGGCAAGGAUUUGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-501-3p | MIMAT0017198 |
Cricetulus griseus | cgr-miR-501-3p | MIMAT0023976 |
Stem-loop Details
Rat
Cricetulus griseus
