Assay Details
Mature miRNA Sequence: | CAUGGUUAGAUCAAGCACAA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-218b | MIMAT0017838 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | rno-miR-218b |
|---|---|
| Stem-loop Accession Number: | MI0015428 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAUGGUUAGAUCAAGCACAA |
| Species: | Rat |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | CAUGGUUAGAUCAAGCACAA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-218b | MIMAT0017838 |