siRNA Details
| Gene Symbol: TSSK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053006.4 | NP_443732.3 | 1 | 1761 | ||
| GenBank mRNA | AF362953.1 | - | 1 | 1241 | ||
| BC037781.2 | - | 1 | 1292 | 5273397 | ||
| L77564.1 | - | 1 | 1761 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSSK2| Gene Symbol: TSSK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053006.4 | NP_443732.3 | 1 | 1761 | ||
| GenBank mRNA | AF362953.1 | - | 1 | 1241 | ||
| BC037781.2 | - | 1 | 1292 | 5273397 | ||
| L77564.1 | - | 1 | 1761 | |||