siRNA Details
| Gene Symbol: ARF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001662.3 | NP_001653.1 | 2 | 249 | ||
| GenBank mRNA | AF493884.1 | - | 2 | 102 | ||
| AK297026.1 | - | 2 | 154 | |||
| AK307297.1 | - | 2 | 223 | |||
| AK308548.1 | - | 2 | 431 | |||
| AK312107.1 | - | 2 | 154 | |||
| BC003043.1 | - | 2 | 139 | 2822973 | ||
| BC033104.1 | - | 2 | 196 | 4862014 | ||
| BT007087.1 | - | 2 | 102 | |||
| CN259625.1 | - | 2 | 266 | |||
| M57567.1 | - | 2 | 156 | |||

ARF5
Genomic Map