siRNA Details
| Gene Symbol: UBE2L6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004223.4 | NP_004214.1 | 3 | 231 | ||
| NM_198183.2 | NP_937826.1 | 3 | 590 | |||
| GenBank mRNA | AF031141.1 | - | 3 | 132 | ||
| AF061736.1 | - | 3 | 182 | |||
| AK093462.1 | - | 3 | 544 | |||
| AK129621.1 | - | 3 | 102 | |||
| AK226165.1 | - | 3 | 285 | |||
| AK315755.1 | - | 3 | 150 | |||
| BC032491.1 | - | 3 | 231 | 5246109 | ||
| CR457126.1 | - | 3 | 135 | |||

UBE2L6
Genomic Map