Learn More
Axygen™ 0.5 mL Conical Screw Cap Tubes: Presterilized
Catalog No. 14222527 Shop All Axygen Products
Change view
Click to view available options
Closure Color:
Assorted
Blue
Clear
Green
Red
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color
14-222-527 Green
14-222-517 Assorted
14-222-561 Yellow
14-222-523 Clear
14-222-532 Red
14-222-521 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-222-527 Supplier Axygen™ Supplier No. SCT050GS
Add to Cart
Add to Cart
Features chemical resistant O-ring
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping of restriction enzymes and other reagents.
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
  • Available in range of colors for easy sample identification
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Green
Closure Material Polypropylene
Capacity (Metric) 0.5 mL
Material Polypropylene
Format Non-looped
Includes O-ring
Packaging 500 Tubes and caps/Pk.; 8 Packs/Cs.
Sterility Sterile
Closure Type Screw Cap
Bottom Shape Conical
Max. RCF 20,000 x g
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov