Learn More
Thermo Scientific™ Amber Screw Cap Micro Tubes
GREENER_CHOICE
Catalog No. 14755212
Change view
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
2 mL
Bottom Shape:
Non-skirted
Skirted
Closure Type:
Attached Cap
Not Included
Screw Cap with O-ring (Unattached)
Fit for Purpose:
PCR Ready
Sterile and PCR Ready
31 product options available for selection
Product selection table with 31 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Bottom Shape Closure Type Fit for Purpose
14-755-212 2 mL Non-skirted Attached Cap Sterile and PCR Ready
14-755-201 0.5 mL Skirted Not Included PCR Ready
14-755-273 0.5 mL Skirted Screw Cap with O-ring (Unattached) PCR Ready
14-755-216 0.5 mL Skirted Attached Cap Sterile and PCR Ready
14-755-202 0.5 mL Skirted Not Included Sterile and PCR Ready
14-755-209 0.5 mL Non-skirted Not Included PCR Ready
14-755-307 0.5 mL Non-skirted Screw Cap with O-ring (Unattached) PCR Ready
14-755-306 0.5 mL Non-skirted Attached Cap Sterile and PCR Ready
14-755-210 0.5 mL Non-skirted Not Included Sterile and PCR Ready
14-755-200 0.5 mL Skirted Not Included PCR Ready
14-755-203 0.5 mL Skirted Not Included Sterile and PCR Ready
14-755-219 1.5 mL Non-skirted Not Included PCR Ready
14-755-214 1.5 mL Non-skirted Screw Cap with O-ring (Unattached) PCR Ready
14-755-207 1.5 mL Non-skirted Attached Cap Sterile and PCR Ready
14-755-220 1.5 mL Non-skirted Not Included Sterile and PCR Ready
14-755-275 1.5 mL Skirted Not Included PCR Ready
14-755-276 1.5 mL Skirted Not Included Sterile and PCR Ready
14-755-218 1.5 mL Non-skirted Not Included PCR Ready
14-755-221 1.5 mL Non-skirted Not Included Sterile and PCR Ready
14-755-281 2 mL Skirted Not Included PCR Ready
14-755-278 2 mL Skirted Screw Cap with O-ring (Unattached) PCR Ready
14-755-227 2 mL Skirted Attached Cap Sterile and PCR Ready
14-755-282 2 mL Skirted Not Included Sterile and PCR Ready
14-755-230 2 mL Non-skirted Not Included PCR Ready
14-755-225 2 mL Non-skirted Screw Cap with O-ring (Unattached) PCR Ready
14-755-231 2 mL Non-skirted Not Included Sterile and PCR Ready
14-755-280 2 mL Skirted Not Included PCR Ready
21-403-205 2 mL Skirted Attached Cap Sterile and PCR Ready
14-755-283 2 mL Skirted Not Included Sterile and PCR Ready
14-755-229 2 mL Non-skirted Not Included PCR Ready
14-755-232 2 mL Skirted Not Included Sterile and PCR Ready
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 31 options available.
31 options
This item is not returnable. View return policy
Catalog No. 14-755-212 Supplier Thermo Scientific™ Supplier No. 3463ANK
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Pure Protection: Prepare, transfer and store samples securely with Thermo Scientific Screw Cap Microtubes. Available in multiple volumes, packaging options and clean claims.
Thermo Scientific™ Screw Cap Micro Tubes robust design and material selection are well suited for work with biologics and offer rich features, including ultra-clear externally threaded tubes and UV protective amber tubes. Options are available with or without a self-standing skirted bottom, non-knurl and knurled tubes for single hand operation, in 0.5mL, 1.5mL and 2.0mL sizes, and the option of multicolored caps, with or without tethered straps. Screw Cap Micro Tube Caps offer a durable design incorporating an O-ring that helps prevent leaks under demanding conditions.
Note: For screw caps, see Cat. No. 3471AS.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Capacity (Metric) 2 mL
Color Amber
Material Polypropylene (Virgin)
Includes NonKnurl
Packaging 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
Temperature Range (Metric) Cold Storage: -80° to +95°C. Liquid Nitrogen: Short term Vapor Phase & Quick Flash Freeze acceptable, not recommmended for LN2 submersion storage
Clean Claims Sterile Electron Beam, SAL 10-6 Free of: Rnase/Dnase, Human and Mouse DNA, Pyrogen, Endotoxin, Bioburden, ATP & PCR Inhibitor
UV Resistance (UV) Ultra violet protection avg. 1.22% transmittance between 220 and 500nm
Binding Property Low Nucleic Acid and Protein Binding & Low Enzyme Inhibition
Max. Torque Manual/Hand Torque: Standard: 0.125 lbsf.in to 2.5 lbsf.in; Metric: 1.412 Ncm to 28.24 Ncm. Automated/Robotic Torque Standard: 1.312 lbsf.in to 2.5 lbsf.in; Metric: 14.83 Ncm to 28.24 Ncm
Sterility Sterile
Graduated No
Closure Type Attached Cap
Fit for Purpose Sterile and PCR Ready
Bottom Shape Non-skirted
Max. RCF 20,000 x g
Product Type Screw-cap Microtube
Show More Show Less
† Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.
Screw cap micro tubes and caps are not recommended for autoclave. Sterile options available.
Not intended to submerge and store in liquid phase LN2.