Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PACSIN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675183
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-183 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-183 Supplier Invitrogen™ Supplier No. PA141564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse, rat, canine, bovine, and chicken based on sequence homology.

PACSIN 2, a 486 AA phosphoprotein is a novel member of the PACSIN family of cytoplasmic adapter proteins. It is an isoform of PACSIN with a CDC15 N-terminal domain, a C-terminal SH3 domain, 3 conserved regions specific to the PACSIN family, and 3 asn-pro-phe (NPF) motifs, which potentially bind to EH domains. This protein is known to display a ubiquitous expression pattern with the highest levels of expression in brain. Reports suggest that PACSIN 2 participates in the organization of the actin cytoskeleton, the regulation of vesicular traffic. It also participates in endocytic internalization and endocytic receptor recycling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PACSIN2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin and 0.05% sodium azide
Gene PACSIN2
Gene Accession No. Q9DDA9, Q9QY17, Q9UNF0, Q9WVE8
Gene Alias AI197433; cytoplasmic phosphoprotein PACSIN2; Pacsin2; pacsin2.S; protein kinase C and casein kinase substrate in neurons 2; protein kinase C and casein kinase substrate in neurons 2 protein; protein kinase C and casein kinase substrate in neurons 2 S homeolog; protein kinase C and casein kinase substrate in neurons protein 2; RP3-437M21.1; SdpII; SdpIIsyndapin II; synaptic dynamin-associated protein II; syndapin 2; Syndapin II; syndapin2; syndapin-2; Syndapin-II; XELAEV_18019736mg; x-PACSIN2
Gene Symbols PACSIN2, pacsin2.S
Host Species Rabbit
Immunogen A portion of amino acids 400-450 of human PACSIN2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11252, 124461, 23970, 398138
Target Species Human, Mouse, Rat, Monkey, Frog
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.