Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS Genie SS Beads

Catalog No. 01804402 Shop All OHAUS Products
Click to view available options
Diameter (Metric):
2.4 mm
3.2 mm
5 mm

Catalog No. 01-804-402

Supplier: OHAUS 83041299

Only null left
Add to Cart
Add to Cart

Bead balls are made of Type 316 stainless steel.

  • For bead beating applications.
  • Use with Bead Genie and Disruptor Genie™.
  • Can be cleaned and reused.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Beads
Diameter (Metric) 3.2 mm
Model Bead Genie
Packaging Quantity 100 Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.