Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ 4 ft. Logic™ Class II, Type B2 Biosafety Cabinet, US Model
SDP

Catalog No. 01812557 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Cabinet
Cabinet, Accessory Package
Cabinet, Accessory Package, Base Stand
Cabinet, Base Stand
Plug Type:
North America (115 V, 15 A)
North America (230 V)
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Plug Type
01-812-557 Cabinet, Accessory Package North America (115 V, 15 A)
01-812-561 Cabinet North America (115 V, 15 A)
01-812-559 Cabinet, Base Stand North America (115 V, 15 A)
01-812-555 Cabinet, Accessory Package, Base Stand North America (115 V, 15 A)
01-812-560 Cabinet North America (230 V)
01-812-558 Cabinet, Base Stand North America (230 V)
01-812-556 Cabinet, Accessory Package North America (230 V)
01-812-554 Cabinet, Accessory Package, Base Stand North America (230 V)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 01-812-557 Supplier Labconco™ Supplier No. 303481002
Add to Cart
Add to Cart
Request a Quote

Total exhaust cabinets that provide personnel, product and environmental protection from hazardous particulates requiring BSL 1-4 containment. These models require a dedicated exhaust system and remote blower. All models are NSF/ANSI 49 listed and maximize safety, performance, and comfort.

Performance Features:

  • Safe Class II airflows (100% exhaust, remote blower required) that provide personnel, product, and environmental protection.
    • Cabinet interior provides ISO Class 5 (Grade A) or better working conditions.
  • Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe downflow – even as HEPA filters load.
    • Optimized airflow minimizes energy use and noise output to industry-leading levels.
  • Logic OS with 5 in. line-of-sight touchscreen display.
    • Access cabinet information such as filter life, alerts and alarms, data logs.
    • Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet.
  • Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work.
  • Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work.
  • Remote Communication Board for direct communication with remote blower and/or building management system featuring dry contact relays, USB data output and RS-232 RJ45 data output.
  • Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach.
  • Full, six-year warranty on all Class II biosafety cabinets.

Optional Configurations:

  • Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals
  • Telescoping base stand with shelf and leveling feet

Specifications

Description 4 ft. Logic Class II, Type B2 Biosafety Cabinet
Certifications/Compliance NSF/ANSI 49, ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2
Chamber Material Stainless Steel (304)
Width (Metric) Interior 123.2 cm
Product Type Class II, Type B2 Bio Safety Cabinet
Model Logic B2
Noise Level 61 dBA
Outlets 2 Duplex
Plug Type North America (115 V, 15 A)
Power Consumption 105 W
Receptacles 2 Double, Left and Right Side
Height (English) 72.7 in
Sash Opening (Metric) 20 cm
Width (English) Exterior 54.4 in.
Volume (Metric) Exhaust 1390 m³/hr.
Display 5 in. touchscreen
Filter Efficiency 99.99%
Filter Type HEPA
Width (Metric) Exterior 138.2 cm
Includes Cabinet, Accessory Package
Lighting UV, LED
Height (Metric) 184.7 cm
Sash Opening (English) 8 in.
Service Fixtures 1
Volume 818 CFM
Warranty 6 Years
Width (English) Interior 48.5 in.
Frequency 60 Hz
Height (English) Interior 25.7 in.
Height (English) Max. Opening 22.5 in.
Height (English) Viewing 27 in.
Height (Metric) Interior 65.3 cm
Height (Metric) Max. Opening 57.2 cm
Height (Metric) Viewing 69 cm
Depth (English) 32 in.
Depth (Metric) 81.3 cm
Voltage 115 V
Energy Consumption (Operating Set Point) 105 W
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.