Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Colored Screw Caps
GREENER_CHOICE

Catalog No. 02681366 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Amber
Assorted
Blue
Green
Natural
Orange
Red
Violet
Yellow
Product Type:
Screw Cap with O-ring
Screw Cap without O-ring
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
02-681-366 Violet Screw Cap with O-ring
02-681-358 Natural Screw Cap with O-ring
02-681-359 Blue Screw Cap with O-ring
02-681-360 Yellow Screw Cap with O-ring
02-681-361 Red Screw Cap with O-ring
02-681-363 Green Screw Cap with O-ring
02-681-364 Orange Screw Cap with O-ring
02-681-367 Amber Screw Cap with O-ring
02-681-368 Assorted Screw Cap with O-ring
02-681-347 Natural Screw Cap without O-ring
02-681-354 Violet Screw Cap without O-ring
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 02-681-366
Add to Cart
Add to Cart

For use with nonsterile skirted- and conical-bottom tubes.

For use with nonsterile skirted- and conical-bottom tubes

  • Feature a frosted writing surface
  • Can be pierced for sample removal
Note: For tubes, see Fisherbrand™ Nonsterile Threaded End Microcentrifuge Tubes without Caps.
TRUSTED_SUSTAINABILITY

Specifications

Includes O-ring
Product Type Screw Cap with O-ring
Autoclavable Autoclavable
Material Polypropylene
Color Violet
Sterility Non-sterile
For Use With (Equipment) Non-sterile skirted and conical bottom tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.