Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,1,1,3,3,3-Hexafluoroacetone hydrate, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Melting Point | 18.0°C to 21.0°C |
Density | 1.5800g/mL |
Boiling Point | 105.0°C to 108.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Quantity | 5 g |
Specific Gravity | 1.58 |
Solubility Information | Solubility in water: soluble. |
Formula Weight | 166.02 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Toxic if swallowed. Fatal in contact with skin. Toxic if inhaled. Causes skin irritation. Causes serious eye damage. Suspected of damaging fertility. Suspected of damaging the unborn child. |
Precautionary Statement | GHS P Statement Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction