Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,3,3,3-Hexamethyldisilazane, HMDS, EMD-Millipore

Catalog No. p-4782889
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass

Chemical Identifiers

CAS 999-97-3
Molecular Formula C6H19NSi2
Molecular Weight (g/mol) 161.395
InChI Key FFUAGWLWBBFQJT-UHFFFAOYSA-N
Synonym Bis(trimethylsilyl)amine, HMDS
PubChem CID 13838
ChEBI CHEBI:85068
IUPAC Name [dimethyl-(trimethylsilylamino)silyl]methane
SMILES C[Si](C)(C)N[Si](C)(C)C

Specifications

Packaging Glass
Explosion Limit 0.8 to 25.9 %(V)
Ignition Point 325°C
Vapor Pressure 20 hPa (20°C)
Viscosity 0.9mm2/sec., kinematic (25°C)
Density Density: 0.774g/cm3 (20°C)
Quantity 25 mL
Formula Weight 161.39
Chemical Name or Material 1,1,1,3,3,3-Hexamethyldisilazane
Recommended Storage Store at 15° to 25°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.