Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,5,5,5-Hexamethyl-3-[(trimethylsilyl)oxy]-3-vinyltrisiloxane 98.0+%, TCI America™
SDP

Catalog No. H161325ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 mL
25 mL

Chemical Identifiers

CAS 5356-84-3
Molecular Formula C11H30O3Si4
Molecular Weight (g/mol) 322.70
MDL Number MFCD00054159
InChI Key CHEFFAKKAFRMHG-UHFFFAOYSA-N
Synonym vinyl tris trimethylsiloxy silane, vinyltris trimethylsiloxy silane, 1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy-3-vinyltrisiloxane, tris trimethylsiloxy vinyl silane, trisiloxane, 3-ethenyl-1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy, 4-ethenyl-2,2,6,6-tetramethyl-4-trimethylsilyl oxy-3,5-dioxa-2,4,6-trisilaheptane
PubChem CID 79316
IUPAC Name 4-ethenyl-2,2,6,6-tetramethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptane
SMILES C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)C=C

Specifications

Color Colorless
Boiling Point 88°C
Quantity 25 mL
Formula Weight 322.70
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,5,5,5-Hexamethyl-3-[(trimethylsilyl)oxy]-3-vinyltrisiloxane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.