Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1-Trifluoro-2,4-pentanedione, 98%

Catalog No. p-4510009
Click to view available options
Quantity:
10 mL
25 mL
100 mL

Catalog No. AC216710100

Supplier: Thermo Scientific Chemicals 216710100

Only null left
Add to Cart

CAS: 367-57-7 | C5H5F3O2 | 154.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-57-7
Molecular Formula C5H5F3O2
Molecular Weight (g/mol) 154.09
MDL Number MFCD00000427
InChI Key SHXHPUAKLCCLDV-UHFFFAOYSA-N
Synonym 1,1,1-trifluoro-2,4-pentanedione, trifluoroacetylacetone, 1,1,1-trifluoroacetylacetone, trifluoroacetyl acetone, acetyl trifluoroacetone, 2,4-pentanedione, 1,1,1-trifluoro, tfpd-h, unii-9n20a8g8sw, alpha,alpha,alpha-trifluoroacetylacetone, a,a,a-trifluoroacetylacetone
PubChem CID 73943
IUPAC Name 1,1,1-trifluoropentane-2,4-dione
SMILES CC(=O)CC(=O)C(F)(F)F

Specifications

Color Orange to Yellow
Density 1.2700g/mL
Boiling Point 105.0°C to 107.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3870 to 1.3900
Linear Formula CH3COCH2COCF3
Quantity 10 mL
Beilstein 01,III,3123
Specific Gravity 1.27
Solubility Information Solubility in water: slightly soluble
Formula Weight 154.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,1,1-Trifluoro-2,4-pentanedione
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 206-698-5
Recommended Storage Product may darken in storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.