Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1-Tris(hydroxymethyl)ethane, 97%

Catalog No. p-7027515
Click to view available options
Quantity:
500 g
2500 g

CAS: 77-85-0 | C5H12O3 | 120.148 g/mol

1,1,1-Tris(hydroxymethyl)ethane is an intermediate in the production of alkyd and polyester resins, powder coating resins, synthetic lubricants based on polyol esters, stabilizers for plastics, plasticizers, and pigment coatings. It is used in some phase change materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1-Tris(hydroxymethyl)ethane is an intermediate in the production of alkyd and polyester resins, powder coating resins, synthetic lubricants based on polyol esters, stabilizers for plastics, plasticizers, and pigment coatings. It is used in some phase change materials.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-85-0
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.148
MDL Number MFCD00004687
InChI Key QXJQHYBHAIHNGG-UHFFFAOYSA-N
Synonym trimethylolethane, 1,1,1-tris hydroxymethyl ethane, trimet, methriol, metriol, pentaglycerine, pentaglycerol, methyltrimethanolmethane, 2-hydroxymethyl-2-methylpropane-1,3-diol, tris hydroxymethyl ethane
PubChem CID 6502
IUPAC Name 2-(hydroxymethyl)-2-methylpropane-1,3-diol
SMILES CC(CO)(CO)CO

Specifications

Melting Point ∼190°C
Boiling Point 137°C (15 mmHg)
Flash Point 160°C (320°F)
Quantity 500 g
Beilstein 1304452
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 120.15
Percent Purity 97%
Chemical Name or Material 1,1,1-Tris(hydroxymethyl)ethane
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-063-9
RTECSNumber TY7137500
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.