Learn More
1,1,1-Tris(hydroxymethyl)ethane, 97%
Description
1,1,1-Tris(hydroxymethyl)ethane is an intermediate in the production of alkyd and polyester resins, powder coating resins, synthetic lubricants based on polyol esters, stabilizers for plastics, plasticizers, and pigment coatings. It is used in some phase change materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,1,1-Tris(hydroxymethyl)ethane is an intermediate in the production of alkyd and polyester resins, powder coating resins, synthetic lubricants based on polyol esters, stabilizers for plastics, plasticizers, and pigment coatings. It is used in some phase change materials.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents, water/ moisture.

Specifications
Specifications
Melting Point | ∼190°C |
Boiling Point | 137°C (15 mmHg) |
Flash Point | 160°C (320°F) |
Quantity | 2500 g |
Beilstein | 1304452 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 120.15 |
Percent Purity | 97% |
Chemical Name or Material | 1,1,1-Tris(hydroxymethyl)ethane |
Safety and Handling
Hazard Category | H315-H319-H335 |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 201-063-9 |
RTECSNumber | TY7137500 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.